Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. p-7119333 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 2639-63-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00048884
InChI Key XAPCMTMQBXLDBB-UHFFFAOYSA-N
Synonym Butyric Acid Hexyl Ester
PubChem CID 17525
ChEBI CHEBI:87559
IUPAC Name hexyl butanoate
SMILES CCCCCCOC(=O)CCC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET4203000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.