Learn More
Proteochem Inc HFBA Ampules (Proteomics Grade), 375-22-4, MFCD00004171, 10x1 mL
Supplier: Proteochem Inc LC620610AMPS
MF: C4 H F7 O2, MW: 214.04, Alternative Names: HFBA; Heptafluorobutanoic acid; Perfluorobutanoic acid; Perfluorobutyric acid; 2,2,3,3,4,4,4-Heptafluorobutanoic acid. Heptafluorobutyric Acid (HFBA) is an ion pairing agent for reverse-phase HPLC separation of peptides or proteins.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.