Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 120 Roll
Lateral flow test roll with a nominal capillary flow time of 120 s/4 cm.
$418.00
Specifications
Filter Type | Membrane Filter |
---|---|
Format | Roll |
Material | Nitrocellulose |
Sterility | Non-sterile |
Flow Rate | 120 sec./4 cm |
Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity & Availability | |||||
SHF1200225
|
MilliporeSigma
SHF1200225 |
2 mil |
Each for $418.00
|
|
|||||
SHF1200425
|
MilliporeSigma
SHF1200425 |
4 mil |
Each for $418.00
|
|
|||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains one 100 m slit roll of backed nitrocellulose membrane with a nominal capillary flow time of 120 seconds/4 cm
- Width: 25 mm
Specifications
Membrane Filter | |
Nitrocellulose | |
120 sec./4 cm | |
Lateral Flow Membrane | |
25 mm |
Roll | |
Non-sterile | |
1 | |
100 m |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title