Learn More
TARGETMOL CHEMICALS INC HIF-2ALPHA AGONIST 2 5MG
Supplier:  TARGETMOL CHEMICALS INC T67831
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. HIF-2(alpha) agonist 2 is an HIF-2(alpha) agonist with an EC50 value of 1.68 uM for HIF-2(alpha). HIF-2(alpha) agonist 2 can promote the stability of the ARNT A/B ring in the 3z structure of HIF-2a-ARNT complex without cytotoxicity to 786-o-Reha-Luc cells. HIF-2(alpha) agonist 2 can be used in the study of oxygen metabolism and is closely related to the occurrence of cancer. purity: 98%
Catalog No. 50-257-6456