Learn More
Chemglass Life Sciences High Flow Manifold System with Hard Connect Fittings, 10L-20L Reactor Capacity, M16 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range
SDP
Supplier:  Chemglass Life Sciences CG1969M01
Encompass_Preferred
High Flow Manifold System with Hard Connect Fittings, 10L-20L Reactor Capacity, M16 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range. The Chemglass high flow manifold system allows for easy and safe connection of your circulator to Chemglass's glass process reactors. Manifold helps alleviate excessive mechanical forces placed on the glass vessel inlet/outlet. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on Stainless Steel Beaded Pipe fitting on one end and Stainless Steel 3/4 Male NPT fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately. CG-1969-M-01 Fits Support Frame CG-1965 and CG-1965-X.
Catalog No. 50-153-1348
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.