Learn More
Ted Pella Inc Hi-Grade Mica, Grade V2, 25 x 25mm (1 x 1"), 20/pk
SDP
Supplier:  Ted Pella Inc 526
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Freshly cleaved muscovite mica surfaces have an outstanding even surface, are optically flat, clear, transparent, scratchless and free from fingerprints. They are very useful in electron microscopy for production of carbon support films, particle imaging, cell growing and thin film coating research. Muscovite mica surface are also suitable as substrates for high resolution atomic force microcopy studies such as DNA, DNA-protein and thin films.
Catalog No. NC9159787