Learn More
World Precision Instrument High Resolution Manual Micromanipulator, 5 micrometer resolution
SDP
Supplier:  World Precision Instrument SYSHS6
Encompass_Preferred
Rock solid motorized micromanipulator
Engineered for stability, and built on a twelve-pound steel plate, this instrument is chosen for high res­o­lu­tion micro-re­cord­ing. HS6 serves equal­ly well as a base for other precision micro-drives. HS6 can be bolted directly to a lab fixture or vibration-free platform. Resolution is extremely high — each gra­dua­tion on its large micrometer barrels indicates just 5 µm movements. Rack and pinion drive, V-shaped guideways and cross roller bearings give sure, re­peat­able move­ments without side play, slipping or stick­ing. All contact parts are milled of hard­ened steel. A flexible ball-joint assembly allows the electrode to be positioned at any angle relative to the x, y or z axes. The entire manipulator tilts forward to 25º allowing rapid coarse ad­just­ment of the elec­trode and allowing cell pen­e­tra­tion along the axis of any of the mi­crome­ters.
Catalog No. 50-253-8498
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.