Learn More
XpressBio HIV-1 Reverse Transcriptase Assay Kit
SDP
Supplier:  XpressBio RT1000
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
XpressBio's 2-hour HIV RT assay for drug screening and a 24-hour assay to detect as little as 1 pg of HIV RT isolated from viral conditioned media.  An assay incubation time of 5-6 hours allows the experiment to be completed in one day and will detect 2 pg of HIV RT.  The RT assay is quantitative, non-radioactive, colorimetric, and can be used to determine if test articles, compounds, or peptides can inhibit HIV RT activity. The assay can also be used to evaluate HIV RT activity in experimental samples.
Catalog No. NC1977987