Learn More
Innovative Research Inc Hmn IgG Fc Frgmnt Aff Pur 5mg
Supplier:  Innovative Research Inc IHUIGGFCAP5MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Human IgG Fc Fragment Affinity Purified 5mg - Human IgG Fc Fragment Affinity Purified from Innovative Research has been purified from human plasma by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Phosphate; 0.2M NaCl; 0.05% Azide; pH 8.0 with a purity of 95% as determined by SDS-PAGE.
Catalog No. 50-274-4306