Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNRNPR Polyclonal Antibody, Proteintech

Description
HNRNPR Polyclonal antibody specifically detects HNRNPR in Human samples. It is validated for Immunohistochemistry, Western BlotSpecifications
HNRNPR | |
Polyclonal | |
Unconjugated | |
O43390 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hnRNP-R; HNRNPR; HNRPR | |
Antigen affinity chromatography | |
RUO | |
10236 | |
-20°C | |
Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?