Learn More
Honeywell™ Eclipse™ 6400 Structural Firefighting Gloves
Catalog No. p-3742340
Click to view available options
Size:
Large
2X-Large
Innovative cuff design is convertible and can be worn as a knit wrist, or in a gauntlet style. DuPont™ Nomex™ fibers provide strength and durability, in addition to a high level of thermal protection.
  • Innovative cuff is convertible and can be worn as either a knit wrist or gauntlet cuff
  • Extra-wide cuff, constructed of 2-ply Nomex™ spandex makes donning easy
  • DuPont™ Nomex™ fibers provide superior strength and durability
  • Anti-slash strips provide an extra band of protection to the palm and first knuckle
  • Gloves provide a high level of thermal protection and are capable of standing up to high heat
  • Unique Swing Thumb™ bends in opposition to the fingertips, mimicking the motions of human hands
  • Crosstech™ Moisture Barrier keeps hands dry and comfortable
  • 360° inner pull tab for additional strength in tough applications
Certifications
NFPA 1971
Specifications
Back Material Elkhide Leather
Certifications/Compliance NFPA 1971
Cuff Material Nomex™ Spandex
Coating Material Uncoated
Color Brown, Dark Blue
Hand Specific
Product Type Structural Firefighting Gloves
Size Large
Glove Cuff Color White
Glove Cuff Style Gauntlet
Includes Crosstech™ Moisture Barrier
Liner Material Self Extinguishing Fleece (SEF)
Palm Type Cowhide Leather
Series GL-6400 Eclipse
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.