Learn More
Omnimed Inc Double Door Horizontal Mount Stainless Steel Narcotic Cabinet (8"H X 16"W X 8"D)
SDP
Supplier:  Omnimed Inc 181502
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Omnimed Horizontal Narcotic Cabinets are constructed of durable, heavy duty stainless steel and meet schedule 2 drug storage requirements. They have strong piano hinged doors and a two door, dual locking system to ensure content security. The outside door has a triple bolt wafer lock which opens to a second door secured by a flat key lock with a heavy gauge locking cam. Three extra keys are included for each unique lock. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right outer door access. Pre-drilled holes that are reinforced with stainless steel mounting discs ensure stable wall attachment. Measuring 8H x 16W x 8D.
Catalog No. 50-164-9544
missing translation for 'QuantityLeft'