Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific Horizontal Standard Midi-Gel Kits

Used with Midi Horizontal System.
Supplier: CBS Scientific MHU402PK
Description
CBS Midi Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.- Platinum electrode cassettes can be easily removed for cleaning or replacement
- High quality, heavy duty construction with vast array of accessory options
- Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
- Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
- MHU-602 Midi-Horizontal runs up to a full microtitre plate (108 samples) per gel
- Tapered baffles allow easy in situ gel formation with or without gel tray
- Run up to 32 samples per gel. Two slots in each gel tray
- Combs are printed with catalog number and thickness for easy identification
- Choice of double-sided adjustable height combs gives researcher more options
- Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
- Horizontal Midi-Gels are available in four vivid colors; green, yellow, blue or fluorescent pink
- Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Specifications
CE | |
Compatible Gel Tray: WGT-400 | |
22.5 x 13 x 14 cm (8.8 x 5.1 x 5.5 in.) | |
L x W: 11 x 10.5 cm (4.3 x 4.1 in.) | |
3 Years |
Pink | |
Gel Tray I.D.: 11 x 9.2 cm (4.3 x 3.6 in.) | |
Midi-Gel Kit | |
550 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction