Learn More
Foxx Life Sciences PUREGRIP® HPLC Solvent Reservoir 38-430 System with Class VI PTFE Adapter & Safety Air Inlet Valve, Cap-Only,One (1x) Ports- OD Tubing-3.2 mm (1/8"") or 1.6 mm (1/16""), 1/EA
Supplier: Foxx Life Sciences 4500302FLS
Foxx Life Sciences PUREGRIP® HPLC Solvent Reservoir Caps & Bottle system offer the safest & most ergonomic lab safety solution in the industry. This system come w/ VersaCap®, a Class VI PTFE Manifold, chemically resistant Viton O-Ring & an Air Inlet Valve combined w/ a Foxx 25mm, 0.2m PTFE Filter for safe vapor filtration.The Air Inlet Valve & PTFE Filter block out all harmful solvent vapors while blocking any incoming contaminants from the incoming air.The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap & eliminates the tubes from sliding in & out of the container.Each assembly comes w/ 38-430 Cap & Manifold Assy., Safety Inlet vale w/ filter, fittings for 1x 3.2mm (1/8") OD Tubing, 1x 1.6mm (1/16") OD Tubing & 1x Plug of 6.4 mm(1/4") for blocking of unused ports.*Filter membrane absorbs contaminants from the flowing air, it is recommended that one replaces the vent filter every 6 months to ensure quality operation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.