Learn More
Foxx Life Sciences EZWaste® HPLC 38-430 Solvent Waste System, Cap-Only W/ Exhaust Filter, Three (3x) Ports for OD Tubing 3.2 mm (1/8"") or 1.6 mm (1/16""), 1/EA
Supplier: Foxx Life Sciences 3300101FLS
Foxx Life Sciences EZwaste® HPLC 38-430 Solvent Waste system is designed to be an affordable solution for HPLC liquid waste disposal. Each system is Manufactured to safely collect waste from multiple HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers & provides safer mechanism of disposal for vapor wastes.The EZwaste® HPLC Solvent Waste system also features a closed loop design which protects against leaks & spills, improving overall lab safety. Each EZwaste® Assy. includes a VersaCap® 38-430, Manifold w/ 3-Ports, 3x fittings of 3.2mm (1/8") OD Tubing, 3x fittings of 1.6mm(1/16") OD Tubing, 3x suitable Plugs for blocking of unused ports & 1 Carbon Exhaust Filter. So EZWaste® HPLC 38-430 Solvent Waste Cap Assy. can be used use w/ all type of HPLC machine.* Carbon exhaust filter life cycle may vary.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.