Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HQL 79, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Orally active and specific inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) (IC50 = 6 μM). Antiallergic and anti-inflammatory; displays antiasthmatic activity and potent antihistaminic properties in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 377.48 |
Percent Purity | >99% |
Chemical Name or Material | HQL 79 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction