Learn More
Wilmad Labglass HT MNTL HEMI 2000ML
Heating Mantle, Hemispherical. Size: 2000mL. Watts: 380. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.