Learn More
Wilmad Labglass HT MNTL HEMI 50ML
Supplier:  Wilmad Labglass LG8800100
Encompass_Preferred
Heating Mantle, Hemispherical. Size: 50mL. Watts: 60. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-159
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.