Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Calcium Channel beta-4 (aa 389-495) Control Fragment Recombinant Protein
SDP

Catalog No. RP90685
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Click to view available options
:

Recombinant Protein

This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Accession Number

O00305

Concentration

≥5.0 mg/mL

For Use With (Application)

Blocking Assay, Control

Formulation

1M urea, PBS with no preservative; pH 7.4

Gene ID (Entrez)

785

Name

Calcium Channel beta-4

Quantity

100 μL

Storage Requirements

-20° C, Avoid Freeze/Thaw Cycles

Regulatory Status

RUO

Gene Alias

3110038O15Rik; CAB4; Cacnb4; CACNLB4; calcium channel beta 4 subunit; calcium channel voltage-dependent subunit beta 4; calcium channel, voltage-dependent, beta 4 subunit; calcium voltage-gated channel auxiliary subunit beta 4; Cchb4; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; EA5; EIG9; EJM; EJM4; EJM6; lh; OTTHUMP00000207247; OTTHUMP00000207249; Voltage-dependent L-type calcium channel subunit beta-4

Gene Symbol

Cacnb4

Conjugate

Unconjugated

Species

Human

Recombinant

Recombinant

Sequence

EHLGEYLEAYWRATHTTSSTPMTPLLGRNLGSTALSPYPTAISGLQSQRMRHSNHSTENSPIERRSLMTSDENYHNERARKSRNRLSSSSQHSRDHYPLVEEDYPDS

Expression System

E. coli

Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.