Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Calreticulin (NP_004334.1) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10 μg
Specifications
Specifications
Gene | Calretinin |
Accession Number | NP_004334.1 |
Concentration | 10 μg (at 0.2 μg/μL) |
Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
Insert Type | 1267 bp |
Gene ID (Entrez) | 794 |
Gene Alias | 29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, calbindin 2, (29 kD, calretinin), calbindin 2, 29 kDa (calretinin), calbindin D29K, calretinin, CR |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Human |
Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction