Learn More
R&D Systems™ VersaClone cDNA, Human CD1b (NP_001755)
Shop All R&D Systems ProductsOrder Info
Ships at ambient temperature
Specifications
Accession Number
NP_001755
Concentration
10 μg at 0.2 μg/μL
Formulation
cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type
1015bp
Content And Storage
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species
Human
Quantity
10 μg
For Reasearch Use Only; Not for Use in Humans
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.