Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human CLEC4E (NP_055173) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10μg
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human CLEC4E (NP_055173) VersaClone cDNA clone contains the complete ORF for CLEC4E, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CLEC4E insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
Quantity | 10μg |
Species | Human |
Gene | CLEC4E |
Accession Number | NP_055173 |
Concentration | 10μg at 0.2μg/μL |
Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.238 |
Insert Type | 672bp |
Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Gene ID (Entrez) | 26253 |
Gene Alias | CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, MINCLE, MINCLEC-type lectin superfamily member 9 |
Show More |
For Reasearch Use Only. Not for use in Humans.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction