Learn More
*HELICA BIOSYSTEMS HUMAN CRP QUANTITATIVE
Supplier:  *HELICA BIOSYSTEMS 961CRP01H96
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Clinical Diagnostic Test Kits; C-Reactive Protein Assay; ELISA Test Kit, Human CRP Quantitative detection, 96-strip microwell plate based assay, Results in 1.5 hours , One-year stability; Excellent reproducibility
Catalog No. 50-980-601
missing translation for 'QuantityLeft'