Learn More
R&D Systems™ Human CTRP3/C1qTNF3/CORS26 (NP_852100) VersaClone cDNA
Shop All R&D Systems ProductsSpecifications
Gene
hC1QTNF3
Accession Number
NP_852100
Concentration
10 μg at 0.2 μg/μL
Formulation
cDNA is provided in 10 mM Tris-Cl
Insert Type
973bp
pH
8.5
Gene ID (Entrez)
114899
Gene Alias
C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26
Content And Storage
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species
Human
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.