Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cu/ZnSOD Human ELISA Kit
SDP

For the quantitative detection of human Cu/ZnSOD.

$656.00 - $4805.00

Specifications

Accession Number P00441
Assay Range 0.08-5.0 ng/mL
Assay Sensitivity 0.04 ng/mL
Conjugate HRP
Product Type ELISA Kit
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cu/ZnSOD
HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
Human Cu/ZnSOD Standard, 5ng/mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Phosphate Buffered Saline Concentrate (PBS) 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-180-57
SDP
Encompass
View Documents
Invitrogen™
BMS222TEN
10 x 96 Tests
Each for $4,805.00
missing translation for 'QuantityLeft'
Add to Cart
 
50-180-54
SDP
Encompass
View Documents
Invitrogen™
BMS222
96 Tests
Each for $656.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

The Human Cu/Zn Superoxide Dismutase(Hu Cu/ZnSOD)) ELISA quantitates Hu Cu/ZnSOD in human serum, plasma, amniotic fluid, urine, fetal umbilical vein blood, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Cu/ZnSOD. Principle of the method The Human Cu/ZnSOD solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

P00441
0.04 ng/mL
ELISA Kit
Human
Colorimetric Microplate Reader
ALS,ALS1,HEL-S-44,IPOA,SOD,hSod1,homodimer
5.1%
HRP
Amniotic Fluid, 10 μL; Fetal Umbilical Vein Blood,10 μL; Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL; Urine,10 μL
Cu/Zn Superoxide Dismutase
1 hr. 20 min.
0.08-5.0 ng/mL
HRP
Amniotic Fluid, Fetal Umbilical Vein Blood, Plasma, Serum, Supernatant, Urine
ELISA
6647
5.8%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
RUO
2°C to 8°C
Human
1 hr. 10 min.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.