Learn More
ACROBiosystems resDetect™ Human Fc Tag DLL4 ELISA KIT (Residue Testing)
Supplier:  ACROBiosystems RESA05296TESTS
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Human Fc Tag DLL4 ELISA KIT is based on ELISA sandwich method and designed to measure human Fc Tag DLL4 levels in cell culture supernates, serum, and plasma. It contains recombinant human Fc Tag DLL4 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human Fc Tag DLL4 (ACROBiosystems, cat#GMP-DL4H28) concentrations. The specificity has been verified.
Catalog No. 50-271-8978