Learn More
Invitrogen™ Human Dopamine beta Hydroxylase (aa 518-600) Control Fragment Recombinant Protein


Description
Specifications
Specifications
Accession Number | P09172 |
Concentration | ≥5.0 mg/mL |
For Use With (Application) | Blocking Assay, Control |
Formulation | 1M urea, PBS with no preservative; pH 7.4 |
Gene ID (Entrez) | 1621 |
Name | Dopamine beta Hydroxylase |
Quantity | 100 μL |
Storage Requirements | -20° C, Avoid Freeze/Thaw Cycles |
Regulatory Status | RUO |
Gene Alias | Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.