Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human Dopamine Transporter (aa 164-234) Control Fragment Recombinant Protein


Click to view available options
:
100 μL, Unconjugated
Description
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications
Specifications
Accession Number | Q01959 |
Concentration | ≥5.0 mg/mL |
For Use With (Application) | Blocking Assay, Control |
Formulation | 1M urea, PBS with no preservative; pH 7.4 |
Gene ID (Entrez) | 6531 |
Name | Dopamine Transporter |
Quantity | 100 μL |
Storage Requirements | -20° C, Avoid Freeze/Thaw Cycles |
Regulatory Status | RUO |
Gene Alias | DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction