Learn More
ACROBiosystems ACROBiosystems Human EGFRvIII-coupled Magnetic Beads
Supplier:  ACROBiosystems MBSK02010MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 44 μg EGFRvIII protein/1 mg beads can bind Anti-EGFR Antibody (Cetuximab) with an EC50 of 0.7172 μg/mL (QC tested).;Immobilized 44 μg EGFRvIII protein/1mg beads can bind Anti-EGFRvIII Antibody with an EC50 of 0.8856 μg/mL (QC tested).
Catalog No. 50-210-7060
missing translation for 'QuantityLeft'