Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Estrogen Receptor alpha (aa 1-21) Synthetic Peptide

Catalog No. PEP036 Shop All Thermo Scientific Products
Click to view available options
:

Synthetic Peptide

Human Estrogen Receptor A (ERa) (residues 1-21) Synthetic Peptide, PEP-036, from Invitrogen.

Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. PEP-036 is a 21 amino acid synthetic peptide whose sequence corresponds to N-terminal amino acid residues 1-21 of human estrogen receptor a. The sequence of this peptide is (amino to carboxy terminus): M1-T-M-T-L-H-T-K-A-S-G-M-A-L-L-H-Q-I-Q-G-N21- (C) (A cysteine residue was added to facilitate coupling KLH).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human estrogen receptor a, catalog &Num; PA1-308. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.

Specifications

Gene ID (Entrez)

2099

Species

Human

Purification Method

purified

Accession Number

P03372

Common Name

Human Estrogen Receptor alpha (aa 1-21)

Concentration

0.5 mg/mL

Content And Storage

-20°C. Avoid freeze-thaw cycles.

Description

Synthetic Peptide

Format

Lyophilized

Formulation

peptide with no preservative

For Use With (Application)

Control

Gene Symbol

ESR1

Lyophilized

Yes

Preservative

No preservative

Product Type

Peptide

Quantity

50 μg

Regulatory Status

RUO - research use only

Sequence

M1 - T - M - T - L - H - T - K - A - S - G - M - A - L - L - H - Q - I - Q - G - N21 - (C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.