Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GDF-8/Myostatin ELISA Kit
SDP

Catalog No. p-7219402
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Click to view available options
Quantity:

Human GDF-8/Myostatin quantitates human GDF-8/Myostatin in serum, plasma, supernatant.

Human GDF-8/Myostatin quantitates human GDF-8/Myostatin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GDF-8/Myostatin.

The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.

Specifications

Accession Number

Cmpt; compact; GDF8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; myostatin

Assay Range

0.83 to 200 ng/mL

Assay Sensitivity

0.83 ng/mL

Conjugate

Biotin

Product Type

ELISA

Sample Type

Plasma, Serum, Supernatant

For Use With (Equipment)

Colorimetric Microplate Reader

Gene ID (Entrez)

O14793

Gene Symbol

Mstn

Interassay CV

<12%

Intraassay CV

<10%

Kit Contents

Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers

Label or Dye

HRP

Quantity

5 x 96 Tests

Regulatory Status

Research Use Only

Sample Volume

Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL

Storage Requirements

2°C to 8°C

Target

Myostatin

Test Time

1 hr 20 min

Total Assay Time

4 hr 45 min

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.