Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Granzyme H (NP_219491) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
Host Species: Human
Description
The Human Granzyme H (NP_219491) VersaClone cDNA clone contains the complete ORF for Granzyme H, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Granzyme H insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
Gene | hGZMH |
Accession Number | NP_219491 |
Concentration | 10 μg at 0.2 μg/μL |
Formulation | cDNA is provided in 10 mM Tris-Cl |
Insert Type | 754bp |
pH | 8.5 |
Gene ID (Entrez) | 2999 |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction