Learn More
ACROBiosystems resDetect™ Human Interleukin-1 beta (IL-1 beta) ELISA Kit (Residue Testing)
Supplier: ACROBiosystems CRSA01296TESTS
Human Interleukin-1 beta (IL-1β) ELISA Kit is based on ELISA sandwich method and designed to measure human IL-1β levels in cell culture supernates, serum, and plasma. It contains recombinant human IL-1β and a pair of antibodies against the recombinant human IL-1β, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human IL-1β (ACROBiosystems, cat#GMP-ILBH16) concentrations. The specificity has been verified.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.