Learn More
ACROBiosystems Human IL-12p70 ELISA Kit
Supplier:  ACROBiosystems CRSB009480TESTS
Encompass_Preferred
HumanInterleukin-12p70 (IL-12p70) OneStep ELISA Kit is a one-step quick immunoassay designed to measure human IL-12p70 levels in cell culture supernates, serum, and plasma. This assay kit is a standard sandwich-ELISA format. It contains recombinant human IL-12p70 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of natural and recombinant human IL-12p70 concentrations. The specificity has been verified.
Catalog No. 50-233-1462
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.