Learn More
ACROBiosystems resDetect™ Human Interleukin-6 (IL-6) ELISA Kit (Residue Testing)
Supplier:  ACROBiosystems CRSA00596TESTS
Encompass_Preferred
Human Interleukin-6 (IL-6) ELISA Kit (Residue Testing) is based on ELISA sandwich method and designed to detect and quantitative determine of GMP human IL-6 in samples from CAR-T product preparation processing. It contains GMP human IL-6 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human IL-6 (ACROBiosystems, cat#GMP-L06H27) concentrations. The specificity has been verified.
Catalog No. 50-271-9692
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.