Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KCNK7 (aa 33-92) Control Fragment Recombinant Protein
SDP

Catalog No. RP108118
Encompass
Click to view available options
:
100 μL, Unconjugated

Recombinant Protein

This gene encodes a member of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel; however, it may require other non-pore-forming proteins for activity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].

Specifications

Accession Number Q9Y2U2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10089
Name KCNK7
Quantity 100 μL
Storage Requirements -20° C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias K2p7.1; KCNK7; Potassium channel subfamily K member 7; potassium channel, subfamily K, member 7; potassium channel, two pore domain subfamily K, member 7; potassium two pore domain channel subfamily K member 7; RGD1565025; TWIK3; two pore domain K+ channel
Gene Symbol KCNK7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence GPPACRLQAELRAELAAFQAEHRACLPPGALEELLGTALATQAHGVSTLGNSSEGRTWDL
Expression System E. coli
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.