Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KCTD14 (aa 2-81) Control Fragment Recombinant Protein
SDP

Catalog No. RP97398
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Click to view available options
:

Recombinant Protein

KCTD14 is a protein coding gene.

Specifications

Accession Number

Q9BQ13

Concentration

≥5.0 mg/mL

For Use With (Application)

Blocking Assay, Control

Formulation

1M urea, PBS with no preservative; pH 7.4

Gene ID (Entrez)

65987

Name

KCTD14

Quantity

100 μL

Storage Requirements

-20° C, Avoid Freeze/Thaw Cycles

Regulatory Status

RUO

Gene Alias

AI449310; BTB/POZ domain-containing protein KCTD14; D7Ertd760e; KCTD14; potassium channel tetramerisation domain containing 14; potassium channel tetramerization domain containing 14

Gene Symbol

KCTD14

Conjugate

Unconjugated

Species

Human

Recombinant

Recombinant

Sequence

WQGCAVERPVGRMTSQTPLPQSPRPRRPTMSTVVELNVGGEFHTTTLGTLRKFPGSKLAEMFSSLAKASTDAEGRFFIDR

Expression System

E. coli

Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.