Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KCTD15 (aa 1-45) Control Fragment Recombinant Protein
SDP

Catalog No. RP99118
Encompass
Click to view available options
:
100 μL, Unconjugated

Recombinant Protein

Childhood and adult obesity in the United States and to a lesser extent the rest of the world has increased dramatically over the past decade. Both environmental and genetic factors are involved in the onset and progression of weight gain. Recently, the potassium channel KCTD15 was identified as a genetic loci associated with higher than normal body mass index (BMI) in humans along with genes such as GNPDA2, MTCH2, FTO, and TMEM18. Further studies on single nucleotide polymorphisms (SNPs) in non-diabetic and diabetic patients showed that FTO was most strongly associated with obesity while MTCH2 and GNPDA2 were still significantly associated with higher than normal BMI levels. At least two isoforms of KCTD15 are known to exist.

Specifications

Accession Number Q96SI1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79047
Name KCTD15
Quantity 100 μL
Storage Requirements -20° C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BC031749; BTB/POZ domain-containing protein KCTD15; Kctd15; potassium channel tetramerisation domain containing 15; potassium channel tetramerization domain containing 15; potassium channel tetramerization domain-containing protein 15
Gene Symbol KCTD15
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence MPHRKERPSGSSLHTHGSTGTAEGGNMSRLSLTRSPVSPLAAQGI
Expression System E. coli
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.