Learn More
Invitrogen™ Human KCTD9 Control Fragment Recombinant Protein

Specifications
Accession Number
Q7L273
Concentration
≥5.0 mg/mL
For Use With (Application)
Blocking Assay, Control
Formulation
1M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez)
54793
Name
KCTD9
Quantity
100 μL
Storage Requirements
-20° C, Avoid Freeze/Thaw Cycles
Regulatory Status
RUO
Gene Alias
BTB/POZ domain-containing protein KCTD9; BTBD27; KCTD9; potassium channel tetramerisation domain containing 9; potassium channel tetramerization domain containing 9
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.