Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Lysyl Oxidase Homolog 3 (NP_115992) VersaClone cDNA

Catalog No. RDC2944 Shop All R&D Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

Specifications

Gene

Lysyl Oxidase Homolog 3/LOXL3

Accession Number

NP_115992

Gene ID (Entrez)

84695

Gene Alias

EC 1.4.3, EC 1.4.3.-, EC 1.4.3.13, LOL3, LOXL, LOXL3, lysyl oxidase homolog 3, lysyl oxidase-like 3, Lysyl oxidase-like protein 3

Content And Storage

Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

Species

Human

Quantity

10 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.