Learn More
ORP150/HSP12A Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF5558SP
Description
ORP150/HSP12A Polyclonal antibody AF5558SP specifically detects ORP150/HSP12A in Human samples. It is validated for Western BlottingSpecifications
ORP150/HSP12A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, HYOU1, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
Goat | |
Affinity Purified | |
RUO | |
10525 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Y4L1 | |
HYOU1 | |
Mouse myeloma cell line NS0-derived recombinant human ORP150/HSP12A Met695-Leu994 Accession # Q9Y4L1 | |
25 μg | |
Primary | |
Detects human ORP150/HSP12A in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) HSP-70, rhHSPA-2, and rhHSPH-1 is observed. | |
Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only