Learn More
ATCC Human parainfluenza; virus 3; Strain: ATCC-2011-5
Supplier:  ATCC VR1782
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Human parainfluenza virus 3 strain ATCC-2011-5 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This virus was isolated in 2011 from a clinical sample from a patient positive for PIV 3 in Virginia. It has applications in respiratory disease research.
Catalog No. 50-238-2269
missing translation for 'QuantityLeft'