Learn More
ACROBiosystems HEK293/Human RAGE Stable Cell Line
Supplier:  ACROBiosystems CHEKATP1562VIALS
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Advanced glycosylation end product-specific receptor (AGER) is also known as RAGE, and is a single-pass type I membrane protein which contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. AGER is highly expressed in endothelial cells. AGER mediates interactions of advanced glycosylation end products (AGE), and also accumulates in vascular tissue in aging and at an accelerated rate in diabetes, as well as acts as a mediator of both acute and chronic vascular inflammation in conditions such as atherosclerosis and in particular as a complication of diabetes. AGER / RAGE signaling plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.
Catalog No. 50-271-9744
missing translation for 'QuantityLeft'