Learn More
ACROBiosystems Human ROR1-coupled Magnetic Beads
Supplier:  ACROBiosystems MBRK0092MG
Encompass_Preferred
Human ROR1 protein is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403(Accession # Q01973-1).
Catalog No. 50-233-0888
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.