Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Mouse anti-Human, Alexa Fluor 488, Clone: 902320, R&D Systems™
Mouse MonoclonalAntibody
Supplier: R&D Systems FAB8367G100UG
This item is not returnable.
View return policy
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry.Specifications
SLC5A5/Sodium Iodide Symporter | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q92911 | |
SLC5A5 | |
NS0 mouse myeloma cell line transfected with human SLC5A5 Accession # Q92911 | |
100 ug | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
Monoclonal | |
Stains human SLC5A5 transfectants but not irrelevant transfectants in flow cytometry. |
Flow Cytometry | |
902320 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NIS, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
Mouse | |
IgG2a | |
RUO | |
Primary | |
6528 | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction