Learn More
SLC5A5/Sodium Iodide Symporter Mouse anti-Human, Alexa Fluor 594, Clone: 902320, R&D Systems™
Mouse MonoclonalAntibody
Supplier:  R&D Systems FAB8367T100UG
Catalog No. FAB8367T100
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
SLC5A5/Sodium Iodide Symporter
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q92911
SLC5A5
NS0 mouse myeloma cell line transfected with human SLC5A5 Accession # Q92911
RUO
6528
Human
IgG2a
Flow Cytometry
902320
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NIS, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Mouse
100 μg
Primary
Stains human SLC5A5 transfectants but not irrelevant transfectants in flow cytometry.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)