Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TAB1 Synthetic Peptide

Catalog No. PEP0202 Shop All Thermo Scientific Products
Click to view available options
:
50μg

Synthetic Peptide

TAB1 Synthetic Peptide, PEP-0202, from Invitrogen.

This peptide corresponds to 13 amino acids near the center of human TAB1.PEP-0202 can be used as a blocking peptide with polyclonal antibody PA5-20084.

Specifications

Gene ID (Entrez) 10454
Species Human
Purification Method purified
Accession Number Q15750
Common Name Human TAB1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol TAB1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.