Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human TGF-beta 3 DuoSet ELISA
Shop All R&D Systems Products
Click to view available options
:
15 96-well plates
Description
The Human TGF-beta 3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 3.
Specifications
Specifications
Conjugate | Biotin |
Assay Range | 31.20 to 2000 pg/mL |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 7043 |
Synonym | ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction