Learn More
Indigo Biosciences Human Transforming Growth Factor beta Receptors I & II (TGFbR) All-inclusive Cell-Based Luciferase Reporter Assay Kit, 96-well assay plate format
Supplier:  Indigo Biosciences IB12001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Human Transforming Growth Factor beta Receptors I & II (TGFbR). INDIGO's TGFbR reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of TGFbR. In addition to TGFbR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against human TGFbR. This kit provides researchers with: Clear, Reproducible Results, Exceptional Cell Viability Post-Thaw, Consistent Results Lot to Lot. Kits must be stored at -80C. Do not store in liquid nitrogen. Note Reporter Cells cannot be refrozen or maintained in extended culture.
Catalog No. 50-223-1824
missing translation for 'QuantityLeft'