Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TMEM175 (aa 160-186) Control Fragment Recombinant Protein
SDP

Catalog No. RP103843
Encompass
Click to view available options
:
100 μL, Unconjugated

Recombinant Protein

TMEM175 gene ontology annotations related to this gene include potassium channel activity.

Specifications

Accession Number Q9BSA9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84286
Name TMEM175
Quantity 100 μL
Storage Requirements -20° C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 3010001K23Rik; AI504381; endosomal/lysomomal potassium channel TMEM175; endosomal/lysosomal potassium channel TMEM175; hTMEM175; mTMEM175; RGD1309712; TMEM175; Transmembrane protein 175; transmembrane protein 175; endosomal/lysomomal potassium channel TMEM175
Gene Symbol TMEM175
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence YAFHFPHLLSPQIQRSAHRALYRRHVL
Expression System E. coli
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.